CH6371381743 Chem Impex International, Inc.

L-Arginine-[4-nitro-3-(carboxy-iso-propyl amid)-phenyl] ester dihydrochloride

  • MDL number MFCD06796879
  • Cas Number : 1272755-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White to light yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-arginine-4-nitro-3-carboxy-iso-propyl-amid-phenyl-ester-dihydrochloride/7482?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-arginine-4-nitro-3-carboxy-iso-propyl-amid-phenyl-ester-dihydrochloride/7482?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart