CH6371381749 Chem Impex International, Inc.

Boc-1,3-cis-diaminomethylcyclohexane HCl

  • MDL number MFCD04973128
  • Cas Number : 1049743-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201-209 °C (dec.)
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-1-3-i-cis-i-diaminomethylcyclohexane-font-size-2-face-symbol-middot-font-hcl/7488?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-1-3-i-cis-i-diaminomethylcyclohexane-font-size-2-face-symbol-middot-font-hcl/7488?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart