CH6371381753 Chem Impex International, Inc.

Nalpha-Boc-Ngamma-(2,4-dinitrophenyl)-L-2,4-diaminobutyric acid

  • MDL number MFCD06796880
  • Cas Number : 1263045-99-3
  • Molecular Weight : 384.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-58 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form Yellow powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-gamma-sup-2-4-dinitrophenyl-l-2-4-diaminobutyric-acid/7492?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-i-i-boc-i-n-i-sup-gamma-sup-2-4-dinitrophenyl-l-2-4-diaminobutyric-acid/7492?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart