CH6371381754 Chem Impex International, Inc.

Nalpha-Boc-Nbeta-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-L-2,3-diaminopropionic acid

  • MDL number MFCD05663730
  • Cas Number : 1263045-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 143-149 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-beta-sup-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-l-2-3-diaminopropionic-acid/7493?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-beta-sup-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-l-2-3-diaminopropionic-acid/7493?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart