CH6371381762 Chem Impex International, Inc.

Nalpha-Boc-Nepsilon-biotinyl-D-lysine

  • MDL number MFCD04973110
  • Cas Number : 1272755-71-1
  • Molecular Weight : 472.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-128 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-biotinyl-d-lysine/7501?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-boc-i-n-i-sup-epsilon-sup-biotinyl-d-lysine/7501?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart