CH6371381772 Chem Impex International, Inc.

Nalpha-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Ngamma-Fmoc-D-2,4-diaminobutyric acid

  • MDL number MFCD05663737
  • Cas Number : 1263046-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-156 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-i-n-i-sup-gamma-sup-fmoc-d-2-4-diaminobutyric-acid/7514?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-ethyl-i-n-i-sup-gamma-sup-fmoc-d-2-4-diaminobutyric-acid/7514?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart