CH6371381778 Chem Impex International, Inc.

Dde-L-Leu-ol

  • MDL number MFCD06796883
  • Cas Number : 1263045-95-9
  • Molecular Weight : 281.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81-88 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/dde-l-leu-ol/7520?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/dde-l-leu-ol/7520?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart