CH6371381787 Chem Impex International, Inc.

H-D-Tpi(For)-OH HCl

  • MDL number MFCD06795843
  • Cas Number : 1423017-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/h-d-tpi-for-oh-font-size-2-face-symbol-middot-font-hcl/7529?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/h-d-tpi-for-oh-font-size-2-face-symbol-middot-font-hcl/7529?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart