CH6371381788 Chem Impex International, Inc.

O-Allyl-D-tyrosine methyl ester hydrochloride

  • MDL number MFCD04973250
  • Cas Number : 367517-26-8
  • Molecular Weight : 271.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 190-196 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-o-i-allyl-d-tyrosine-methyl-ester-hydrochloride/7530?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-o-i-allyl-d-tyrosine-methyl-ester-hydrochloride/7530?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart