CH6371381791 Chem Impex International, Inc.

Fmoc-cis-1,2-aminocyclohex-4-ene carboxylic acid

  • MDL number MFCD02682628
  • Cas Number : 1212305-88-8
  • Molecular Weight : 363.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-143 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-i-cis-i-1-2-aminocyclohex-4-ene-carboxylic-acid/7533?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-i-cis-i-1-2-aminocyclohex-4-ene-carboxylic-acid/7533?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart