CH6371381793 Chem Impex International, Inc.

Fmoc-1,4-trans-diaminocyclohexane hydrochloride

  • MDL number MFCD03001746
  • Cas Number : 672310-17-7
  • Molecular Weight : 372.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 160 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-1-4-i-trans-i-diaminocyclohexane-hydrochloride/7537?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-1-4-i-trans-i-diaminocyclohexane-hydrochloride/7537?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart