CH6371381797 Chem Impex International, Inc.

(S)-Nalpha-Fmoc-Nbeta-Boc-2,3-diaminopropan-1-ol

  • MDL number MFCD04973190
  • Cas Number : 915714-21-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 149-159 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-beta-sup-boc-2-3-diaminopropan-1-ol/7541?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-beta-sup-boc-2-3-diaminopropan-1-ol/7541?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart