CH6371381800 Chem Impex International, Inc.

Nalpha-Fmoc-Ngamma-(2,4-dinitrophenyl)-D-2,4-diaminobutyric acid

  • MDL number MFCD06796890
  • Cas Number : 1263047-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-120 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form Yellow crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-gamma-sup-2-4-dinitrophenyl-d-2-4-diaminobutyric-acid/7545?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-gamma-sup-2-4-dinitrophenyl-d-2-4-diaminobutyric-acid/7545?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart