CH6371381801 Chem Impex International, Inc.

Nalpha,gamma-Bis-Fmoc-D-2,4-diaminobutyric acid

  • MDL number MFCD02094099
  • Cas Number : 1217645-10-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 183-190 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-g-sup-bis-fmoc-d-2-4-diaminobutyric-acid/7546?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-a-g-sup-bis-fmoc-d-2-4-diaminobutyric-acid/7546?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart