CH6371381804 Chem Impex International, Inc.

Nalpha-Fmoc-Nbeta-4-methyltrityl-D-2,3-diaminopropionic acid

  • MDL number MFCD06796892
  • Cas Number : 1263046-35-0
  • Molecular Weight : 582.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point > 110 °C (dec.)
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-beta-sup-4-methyltrityl-d-2-3-diaminopropionic-acid/7549?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-beta-sup-4-methyltrityl-d-2-3-diaminopropionic-acid/7549?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart