CH6371381806 Chem Impex International, Inc.

Nalpha-Fmoc-Nepsilon-biotinyl-D-lysine

  • MDL number MFCD00270742
  • Cas Number : 110990-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-186 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-biotinyl-d-lysine/7551?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-biotinyl-d-lysine/7551?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart