CH6371381807 Chem Impex International, Inc.

Nalpha-Fmoc-Nepsilon-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)-3-methylbutyl-D-lysine

  • MDL number MFCD06796894
  • Cas Number : 1272755-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White to off-white crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-1-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-3-methylbutyl-d-lysine/7552?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-fmoc-i-n-i-sup-epsilon-sup-1-4-4-dimethyl-2-6-dioxocyclohex-1-ylidene-3-methylbutyl-d-lysine/7552?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart