CH6371381809 Chem Impex International, Inc.

Fmoc-D-methionine sulfone

  • MDL number MFCD06796896
  • Cas Number : 1247791-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-160 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-d-methionine-sulfone/7554?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-d-methionine-sulfone/7554?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart