CH6371381812 Chem Impex International, Inc.

N,N'-Di-Boc-N-Z-hydrazine

  • MDL number MFCD04973127
  • Cas Number : 202980-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-78 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-i-n-i-di-boc-n-z-hydrazine/7558?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-i-n-i-di-boc-n-z-hydrazine/7558?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart