CH6371381815 Chem Impex International, Inc.

N-Benzyl-N-methyl-D-methionine methyl ester hydrochloride

  • MDL number MFCD04973100
  • Cas Number : 1272755-11-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-114 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-benzyl-i-n-i-methyl-d-methionine-methyl-ester-hydrochloride/7561?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-benzyl-i-n-i-methyl-d-methionine-methyl-ester-hydrochloride/7561?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart