CH6371381816 Chem Impex International, Inc.

Nalpha-Benzyl-Nalpha-methyl-D-phenylalanine methyl ester hydrochloride

  • MDL number MFCD04973101
  • Cas Number : 1272755-67-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135-141 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-i-sup-alpha-i-sup-benzyl-i-n-i-i-sup-alpha-i-sup-i-i-methyl-d-phenylalanine-methyl-ester-hydrochloride/7562?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-i-sup-alpha-i-sup-benzyl-i-n-i-i-sup-alpha-i-sup-i-i-methyl-d-phenylalanine-methyl-ester-hydrochloride/7562?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart