CH6371381820 Chem Impex International, Inc.

Nalpha-Benzyl-Nalpha-methyl-L-valine methyl ester hydrochloride

  • MDL number MFCD04973102
  • Cas Number : 1272755-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 102-108 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-i-sup-alpha-i-sup-benzyl-i-n-i-i-sup-alpha-i-sup-methyl-l-valine-methyl-ester-hydrochloride/7566?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-i-sup-alpha-i-sup-benzyl-i-n-i-i-sup-alpha-i-sup-methyl-l-valine-methyl-ester-hydrochloride/7566?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart