CH6371381821 Chem Impex International, Inc.

N-Boc,N'-benzyl-1,2-ethylenediamine hydrochloride

  • MDL number MFCD06796898
  • Cas Number : 126402-64-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164-172 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-boc-i-n-i-benzyl-1-2-ethylenediamine-hydrochloride/7567?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-boc-i-n-i-benzyl-1-2-ethylenediamine-hydrochloride/7567?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart