CH6371381822 Chem Impex International, Inc.

N-Boc-N '-ethyl-1,2-ethylene diamine hydrochloride

  • MDL number MFCD06796899
  • Cas Number : 1073659-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-144 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-boc-i-n-i-ethyl-1-2-ethylene-diamine-hydrochloride/7568?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-boc-i-n-i-ethyl-1-2-ethylene-diamine-hydrochloride/7568?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart