CH6371381828 Chem Impex International, Inc.

N-Benzyl-D-phenylalanine methyl ester hydrochloride

  • MDL number MFCD03788629
  • Cas Number : 84028-90-0
  • Molecular Weight : 305.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 141-147 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-benzyl-d-phenylalanine-methyl-ester-hydrochloride/7574?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-benzyl-d-phenylalanine-methyl-ester-hydrochloride/7574?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart