CH6371381830 Chem Impex International, Inc.

Benzyl-L-leucine methyl ester hydrochloride

  • MDL number MFCD04973106
  • Cas Number : 474430-95-0
  • Molecular Weight : 271.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 155-161 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/benzyl-l-leucine-methyl-ester-hydrochloride/7576?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/benzyl-l-leucine-methyl-ester-hydrochloride/7576?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart