CH6371381831 Chem Impex International, Inc.

Nin-Formyl-L-1,2,3,4-tetrahydro-norharmane-3-carboxylic acid hydrochloride

  • MDL number MFCD06796901
  • Cas Number : 1423017-83-7
  • Molecular Weight : 280.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form Light brown powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-in-sup-formyl-l-1-2-3-4-tetrahydro-norharmane-3-carboxylic-acid-hydrochloride/7577?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-in-sup-formyl-l-1-2-3-4-tetrahydro-norharmane-3-carboxylic-acid-hydrochloride/7577?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart