CH6371381840 Chem Impex International, Inc.

Z-cis-3-aminocyclohexanecarboxylic acid

  • MDL number MFCD06796902
  • Cas Number : 952616-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-115 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-cis-i-3-aminocyclohexanecarboxylic-acid/7586?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-cis-i-3-aminocyclohexanecarboxylic-acid/7586?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart