CH6371381841 Chem Impex International, Inc.

Z-cis-4-aminocyclohexane acetic acid

  • MDL number MFCD06796903
  • Cas Number : 75695-14-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 113-120 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-cis-i-4-aminocyclohexane-acetic-acid/7587?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-cis-i-4-aminocyclohexane-acetic-acid/7587?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart