CH6371381844 Chem Impex International, Inc.

1-(Z-amino)-cyclobutyl-1-carboxylic acid

  • MDL number MFCD03426163
  • Cas Number : 190004-53-6
  • Molecular Weight : 249.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-94 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-z-amino-cyclobutyl-1-carboxylic-acid/7590?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-z-amino-cyclobutyl-1-carboxylic-acid/7590?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart