CH6371381845 Chem Impex International, Inc.

Z-L-aspartic acid beta-allyl ester

  • MDL number MFCD06796904
  • Cas Number : 99793-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-beta-allyl-ester/7591?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-l-aspartic-acid-beta-allyl-ester/7591?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart