CH6371381850 Chem Impex International, Inc.

Nalpha-Z-Nbeta-xanthyl-D-asparagine

  • MDL number MFCD06796910
  • Cas Number : 1313054-82-8
  • Molecular Weight : 446.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 181-187 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-beta-sup-xanthyl-d-asparagine/7596?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-beta-sup-xanthyl-d-asparagine/7596?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart