CH6371381853 Chem Impex International, Inc.

(R)-4-Amino-3-(Z-amino)butyl methyl ester hydrochloride

  • MDL number MFCD03788649
  • Cas Number : 845909-53-7
  • Molecular Weight : 302.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 111-117 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-4-amino-3-z-amino-butyl-methyl-ester-hydrochloride/7599?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-4-amino-3-z-amino-butyl-methyl-ester-hydrochloride/7599?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart