CH6371381855 Chem Impex International, Inc.

Nalpha-Z-Ngamma-Fmoc-D-2,4-diaminobutyric acid

  • MDL number MFCD06796914
  • Cas Number : 369611-58-5
  • Molecular Weight : 474.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139-145 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-gamma-sup-fmoc-d-2-4-diaminobutyric-acid/7601?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-z-i-n-i-sup-gamma-sup-fmoc-d-2-4-diaminobutyric-acid/7601?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart