CH6371381860 Chem Impex International, Inc.

1-(Boc-aminomethyl)cyclopropyl-1-carboxylic acid

  • MDL number MFCD04973108
  • Cas Number : 204376-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 121-129 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/1-boc-aminomethyl-cyclopropyl-1-carboxylic-acid/7606?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/1-boc-aminomethyl-cyclopropyl-1-carboxylic-acid/7606?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart