CH6371381862 Chem Impex International, Inc.

Fmoc-1,3-cis-diaminomethyl-cyclohexane hydrochloride

  • MDL number MFCD04973195
  • Cas Number : 1274891-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-117 °C
Purity 99
Storage temp. Store at 0-8 °C
Physical Form White crystalline powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-1-3-i-cis-i-diaminomethyl-cyclohexane-hydrochloride/7608?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-1-3-i-cis-i-diaminomethyl-cyclohexane-hydrochloride/7608?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart