CH6371381868 Chem Impex International, Inc.

L-Norvaline ethyl ester hydrochloride

  • MDL number MFCD08458628
  • Cas Number : 40918-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-119 °C
Purity 99
Storage temp. Store at 0-8°C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/l-norvaline-ethyl-ester-hydrochloride/7614?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/l-norvaline-ethyl-ester-hydrochloride/7614?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart