CH6371381878 Chem Impex International, Inc.

N-Methyl-D-phenylglycine

  • MDL number MFCD08458661
  • Cas Number : 30925-14-5
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form Off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-d-phenylglycine/7628?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-d-phenylglycine/7628?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart