CH6371381879 Chem Impex International, Inc.

N-Methyl-L-phenylglycine

  • MDL number MFCD00038798
  • Cas Number : 2611-88-3
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0-8°C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-phenylglycine/7629?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-methyl-l-phenylglycine/7629?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart