CH6371381880 Chem Impex International, Inc.

Nalpha-Methyl-L-tyrosine methyl ester HCl

  • MDL number MFCD08458662
  • Cas Number : 70963-39-2
  • Molecular Weight : 245.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Store at 0-8 °C
Physical Form Off-white crystal
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-methyl-l-tyrosine-methyl-ester-font-size-2-face-symbol-middot-font-hcl/7630?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-n-i-sup-alpha-sup-methyl-l-tyrosine-methyl-ester-font-size-2-face-symbol-middot-font-hcl/7630?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart