CH6371381885 Chem Impex International, Inc.

Z-cis-D-4-Hydroxyproline

  • MDL number MFCD03428474
  • Cas Number : 130930-25-5
  • Molecular Weight : 265.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/z-i-cis-i-d-4-hydroxyproline/7638?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/z-i-cis-i-d-4-hydroxyproline/7638?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart