CH6371381918 Chem Impex International, Inc.

Fmoc-L-Phenylglycinol

  • MDL number MFCD01317845
  • Cas Number : 215178-44-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/fmoc-l-phenylglycinol/7691?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/fmoc-l-phenylglycinol/7691?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart