CH6371381927 Chem Impex International, Inc.

cis-3-Iodoacrylic acid

  • MDL number MFCD00274189
  • Cas Number : 6214-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Store at 0 - 8 °C
Physical Form White to off-white powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-cis-i-3-iodoacrylic-acid/7703?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-cis-i-3-iodoacrylic-acid/7703?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart