CH6371381942 Chem Impex International, Inc.

Trityl-L-serine lactone

  • MDL number MFCD00270148
  • Cas Number : 88109-06-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-183 °C
Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/trityl-l-serine-lactone/7720?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/trityl-l-serine-lactone/7720?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart