CH6371381959 Chem Impex International, Inc.

(S)-(+)-1-Amino-1-phenyl-2-methoxyethane

  • MDL number MFCD01862273
  • Cas Number : 91298-74-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8°C
Physical Form Colorless liquid
Shipping Temperature RT
Country of Origin USA
Boiling point 105 - 111 °C (10mmHg)
Product Link http://www.chemimpex.com/product/productinfo/i-s-i-1-amino-1-phenyl-2-methoxyethane/7740?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-s-i-1-amino-1-phenyl-2-methoxyethane/7740?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart