CH6371381971 Chem Impex International, Inc.

(3S)-Trichloromethyl-cis-tetrahydropyrrolo[1,2-c]oxazol-1-one

  • MDL number MFCD08274465
  • Cas Number : 1330286-50-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108-112 °C (dec)
Purity 97
Storage temp. Store at 0-8 °C
Physical Form Off-white solid
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/3-i-s-i-trichloromethyl-i-cis-i-tetrahydropyrrolo-1-2-c-oxazol-1-one/7760?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/3-i-s-i-trichloromethyl-i-cis-i-tetrahydropyrrolo-1-2-c-oxazol-1-one/7760?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart