CH6371381978 Chem Impex International, Inc.

Boc-D-Tryptophanol

  • MDL number MFCD00270221
  • Cas Number : 158932-00-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-123
Synonym Boc-(R)-2-amino-3-(3-indolyl)-1-propanol
Purity 99
Storage temp. Store at 0-8
Physical Form White solid
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-d-tryptophanol/7768?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-d-tryptophanol/7768?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart