CH6371381993 Chem Impex International, Inc.

(R)-3-Thienylglycine

  • MDL number MFCD00079614
  • Cas Number : 1194-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym H-D-(3-Thienyl)gly-OH
Purity 0.98
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/i-r-i-3-thienylglycine/7783?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/i-r-i-3-thienylglycine/7783?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart