CH6371382000 Chem Impex International, Inc.

Boc-(S)-3-thienylglycine

  • MDL number MFCD02682479
  • Cas Number : 910309-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118-124
Synonym Boc-L-(3-thienyl)glycine
Purity 99
Storage temp. Store at 0-8
Physical Form White powder
Shipping Temperature Shipped at ambient temperature
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-thienylglycine/7790?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/boc-i-s-i-3-thienylglycine/7790?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart