CH6371382008 Chem Impex International, Inc.

alpha-Methyl-D-4-Fluorophenylalanine

  • MDL number MFCD08061612
  • Cas Number : 422568-68-1
  • Molecular Weight : 197.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Store at 0-8 °C
Physical Form White powder
Shipping Temperature RT
Country of Origin USA
Product Link http://www.chemimpex.com/product/productinfo/alpha-methyl-d-4-fluorophenylalanine/7876?cid=0

Safety Information

MSDS http://www.chemimpex.com/product/productinfo/alpha-methyl-d-4-fluorophenylalanine/7876?cid=0
Availability details for
Quantity
  • Availability
  • Add to Cart